Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-Fenpyroximate | 134098-61-6 | MFCD00274596 | 99.8% | 1 ML
SDP

Catalog No. 5000407663
Encompass_Preferred

(E)-Fenpyroximate is a potent acaricide that functions as a mitochondrial NADH:ubiquinone reductase inhibitor. This compound is widely used as an acaricide, miticide, and insecticide, and is registered for various plant commodities. It is also approved in the EU as an active substance and is currently being investigated for malaria control.

  • Potent acaricide
  • Functions as a mitochondrial NADH:ubiquinone reductase inhibitor
  • Used as a miticide and insecticide
  • Registered for use on various plant commodities
  • Approved in the EU as an active substance
  • Investigated for malaria control
  • Appearance as a white to off-white solid
  • Molecular formula C24H27N3O4

Catalog No. 50-004-07663 Supplier Medchemexpress LLC Supplier No. HYB082510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-Fenpyroximate is a potent acaricide that functions as a mitochondrial NADH:ubiquinone reductase inhibitor. This compound is widely used as an acaricide, miticide, and insecticide, and is registered for various plant commodities. It is also approved in the EU as an active substance and is currently being investigated for malaria control.

  • Potent acaricide
  • Functions as a mitochondrial NADH:ubiquinone reductase inhibitor
  • Used as a miticide and insecticide
  • Registered for use on various plant commodities
  • Approved in the EU as an active substance
  • Investigated for malaria control
  • Appearance as a white to off-white solid
  • Molecular formula C24H27N3O4

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.