Learn More
FUJIFILM Biosciences E-Ferimzone Standard
Catalog No. 5000450352
Encompass_Preferred
E-Ferimzone Standard is a high-purity analytical reference material intended for pesticide residue analysis It is a white to slightly yellowish brown crystalline powder highly pure 99 0% by both HPLC and qNMR This compound is designed for use in research and analytical applications particularly those involving high-performance liquid chromatography HPLC and quantitative NMR techniques Not suitable for food drug or household use
Catalog No. 50-004-50352 Supplier FUJIFILM Biosciences Supplier No. 06304131
E-Ferimzone Standard is a high-purity analytical reference material intended for pesticide residue analysis It is a white to slightly yellowish brown crystalline powder highly pure 99 0% by both HPLC and qNMR This compound is designed for use in research and analytical applications particularly those involving high-performance liquid chromatography HPLC and quantitative NMR techniques Not suitable for food drug or household use