Learn More
Medchemexpress LLC (E)-Ferulic acid | 537-98-4 | 99.9% | 194.19 | 1 ML
SDP
Catalog No. 5000427990
Encompass_Preferred
(E)-Ferulic acid is an isomer of ferulic acid, an aromatic compound found in plant cell walls. It causes phosphorylation of β-catenin, leading to proteasome degradation, increasing the expression of pro-apoptotic factor Bax, and reducing the expression of the survival factor survivin. It can effectively remove reactive oxygen species (ROS) and inhibit lipid peroxidation, and exerts antiproliferative and antimigratory effects in the human lung cancer cell line H1299.
  • Removes reactive oxygen species (ROS)
  • Inhibits lipid peroxidation
  • Exerts antiproliferative effects in human lung cancer cells
  • Exerts antimigratory effects in human lung cancer cells
  • Induces phosphorylation and proteasome degradation of β-catenin
  • Increases expression of pro-apoptotic factor Bax
  • Reduces expression of the survival factor survivin
  • Reduces liver and kidney toxicity and oxidative stress
Catalog No. 50-004-27990 Supplier Medchemexpress LLC Supplier No. HYN0060B10MM/1ML
(E)-Ferulic acid is an isomer of ferulic acid, an aromatic compound found in plant cell walls. It causes phosphorylation of β-catenin, leading to proteasome degradation, increasing the expression of pro-apoptotic factor Bax, and reducing the expression of the survival factor survivin. It can effectively remove reactive oxygen species (ROS) and inhibit lipid peroxidation, and exerts antiproliferative and antimigratory effects in the human lung cancer cell line H1299.
  • Removes reactive oxygen species (ROS)
  • Inhibits lipid peroxidation
  • Exerts antiproliferative effects in human lung cancer cells
  • Exerts antimigratory effects in human lung cancer cells
  • Induces phosphorylation and proteasome degradation of β-catenin
  • Increases expression of pro-apoptotic factor Bax
  • Reduces expression of the survival factor survivin
  • Reduces liver and kidney toxicity and oxidative stress