Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ E-Gel™ 48 Agarose Gels
GREENER_CHOICE

Catalog No. g820802
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 x 8 Gels
8 Gels
Separation Range:
10 to 400 bp
10 to 500 bp
400 to 10,000 bp
50 to 3000 bp
Gel Percentage:
1%
2%
4%
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Separation Range Gel Percentage
G820802 8 Gels 50 to 3000 bp 2%
G800801 8 Gels 400 to 10,000 bp 1%
G820801 8 Gels 400 to 10,000 bp 1%
G820841 4 x 8 Gels 400 to 10,000 bp 1%
G800802 8 Gels 50 to 3000 bp 2%
G820842 4 x 8 Gels 50 to 3000 bp 2%
G800804 8 Gels 10 to 400 bp 4%
G820804 8 Gels 10 to 500 bp 4%
G820844 4 x 8 Gels 10 to 500 bp 4%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. G820802 Supplier Invitrogen™ Supplier No. G820802
Only null left
Add to Cart
Edge
Add to Cart

Invitrogen E-Gel 48 high-throughput precast agarose gels with or without SYBR Safe DNA Gel Stain are designed for fast, convenient, and less-hazardous DNA sample electrophoresis.

Invitrogen E-Gel 48 high-throughput precast agarose gels with or without SYBR Safe DNA Gel Stain are designed for fast, convenient, and less-hazardous DNA sample electrophoresis. Each E-Gel agarose gel cassette contains all components required for efficient gel separation and analysis—just load your samples and run.

Each gel contains 48 sample wells and 4 marker wells in 1%, 2%, or 4% high-resolution agarose with a 3.2-cm run length. The various agarose percentages are used to separate DNA fragments of different sizes:

  • 1% agarose gels: DNA fragments between 400 bp and 10 kb
  • 2% agarose gels: DNA fragments between 50 bp and 3 kb
  • 4% agarose gels: DNA fragments between 10 bp and 500 bp

Each cassette is individually labeled with an EAN39 bar code that is recognized by most commercially available robotic bar code readers.

Features of E-Gel gels:

  • High-throughput analysis—contains 48 sample lanes and 4 marker lanes per gel
  • Automation compatible—suitable for use with multi-channel pipettors and commonly used 8- and 12-pin liquid handling robots
  • Scalable—resolve up to 192 samples in one run by combining up to four E-Base Mother and E-Base Daughter device units
  • Less hazardous choice—contains SYBR Safe DNA stain, a less hazardous alternative to ethidium bromide
  • High performance—detect 3 ng of sample DNA with separation time as little as ∼20 minutes

Less hazardous option

Choose gels with SYBR Safe DNA stain to minimize exposure to hazardous and mutagenic factors such as ethidium bromide and UV. Precast E-Gel agarose gels with SYBR Safe stain, combined with a blue-light trans-illumination, offer better sample integrity and a less hazardous work environment. Unlike ethidium bromide, the DNA stain most commonly incorporated into agarose gels, SYBR Safe DNA Gel Stain is a non-toxic, non-mutagenic DNA stain that is less hazardous for the user, less hazardous for the environment, and can save institutional overhead costs for disposal.

Fast and convenient analysis

E-Gel agarose gels enable fast and high-performance analysis with a detection sensitivity of 3 ng sample DNA per band and a separation time three times faster than that of conventional pour-your-own gels. Each E-Gel agarose gel contains agarose, electrodes, SYBR Safe DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not require any gel or buffer preparation or gel staining steps. Just load your samples and run.

Running and imaging devices

High-throughput E-Gel agarose gels require the Invitrogen E-Base Electrophoresis Device for gel running.

Available in a variety of gel formats

E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The high-throughput E-Gel 96 and 48 agarose gels with SYBR Safe stain are available in 1%, 2%, and 4% gel percentages.

TRUSTED_SUSTAINABILITY

Specifications

Compatible Ladders E-Gel™ 1 Kb Plus Express DNA Ladder
Content And Storage • Each box contains 8 gels

Store strictly at room temperature. Do not freeze.
Avoid cold spots near freezers or other uncontrolled environments.
Gel Percentage 2%
Gel Type E-Gel
Label or Dye SYBR Safe
Separation Range 50 to 3000 bp
Wells 48 + 4-well
Quantity 8 Gels
Shipping Condition Room Temperature
Product Type Agarose Gel

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.