Learn More
Invitrogen™ E-Gel™ High-Throughput Mix Pack
Description
The E-Gel High-Throughput Mix Pack allows researchers to try all of our high-throughput format E-Gel agarose gels. These gels use SYBR safe as a DNA stain to minimize exposure to hazardous and mutagenic factors such as ethidium bromide or UV light. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.
Benefits of using E-Gel agarose gels with SYBR Safe include:
- Better sample integrity – blue-light excitation wavelength minimizes DNA damage, as opposed to UV light
- Safer choice – contains SYBR Safe DNA stain, a safer alternative to ethidium bromide
- High performance – detect 3 ng of sample DNA with a separation time as short as 12 minutes
- No buffers – E-Gel agarose gels do not require buffers to run
The box contains:
- E-Gel 48 Agarose Gels with SYBR Safe DNA Gel Stain, 1%
- E-Gel 48 Agarose Gels with SYBR Safe DNA Gel Stain, 2%
- E-Gel 96 Agarose Gels with SYBR Safe DNA Gel Stain, 1%
- E-Gel 96 Agarose Gels with SYBR Safe DNA Gel Stain, 2%
Running and imaging devices
High-throughput E-Gel agarose gels with SYBR Safe require the E-Gel Power Snap Plus Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Specifications
Specifications
| Format | Precast Gel |
| Gel Type | E-Gel |
| Separation of | DNA |
| Separation Range | 100 to 2000 bp |
| Separation Type | Electrophoresis |
| Quantity | 8 Gels |
| Sterility | Non-sterile |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.