Learn More
Invitrogen™ E-Gel™ with SYBR™ Safe Mix Pack
Description
The E-Gel with SYBR Safe Mix Pack allows researchers to try all standard format E-Gel agarose gels that use SYBR safe as a DNA stain in your workflow. SYBR Safe-stained gels minimize exposure to hazardous and mutagenic factors such as ethidium bromide or UV light. Precast E-Gel agarose gels with SYBR Safe, in combination with blue-light trans-illumination, offer better sample integrity and a safer work environment.
Benefits of using E-Gel agarose gels with SYBR Safe include:
- Better sample integrity – exploits blue-light excitation wavelength to minimize DNA damage, as opposed to UV light
- Safer choice – contains SYBR Safe DNA stain, a safer alternative to ethidium bromide
- High performance – detect 3 ng of sample DNA with a separation time as short as 13 minutes
- Easy sample access – openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
- No buffers – E-Gel agarose gels do not require buffers to run
The box contains:
- E-Gel Agarose Gels with SYBR Safe DNA Gel Stain, 1%
- E-Gel Agarose Gels with SYBR Safe DNA Gel Stain, 2%
- E-Gel CloneWell II Agarose Gels with SYBR Safe, 0.8%
- E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1%
- E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2%
Running and imaging devices
E-Gel agarose gels with SYBR Safe require the E-Gel Power Snap or E-Gel Power Snap Plus electrophoresis systems for gel running and viewing. The systems consist of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.
Specifications
Specifications
| Format | Precast Gel |
| Gel Percentage | 0.8 to 4% |
| Gel Type | E-Gel |
| Separation of | DNA |
| Separation Type | Electrophoresis |
| Wells | 11-well |
| Quantity | 10 Gels |
| Sterility | Non-sterile |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.