Learn More
Medchemexpress LLC (-)-(E)-Guggulsterone | 39025-24-6 | 1 ML
SDP
Catalog No. 5000418894
Encompass_Preferred
(-)-(E)-Guggulsterone is an orally active natural stereoisomer that acts as an antagonist for the Farnesoid X Receptor (FXR). It exhibits potent hypolipidemic properties and suppresses dengue virus (DENV) replication by upregulating antiviral interferon responses. This compound also possesses antibacterial activities against Bacillus subtilis, Staphylococcus aureus, and Pseudomonas aeruginosa, and has cardiac protective and antioxidant activities.
  • Acts as an antagonist for Farnesoid X Receptor (FXR)
  • Exhibits potent hypolipidemic properties
  • Suppresses dengue virus (DENV) replication
  • Possesses antibacterial activities against Bacillus subtilis, Staphylococcus aureus, and Pseudomonas aeruginosa
  • Has cardiac protective and antioxidant activities
Catalog No. 50-004-18894 Supplier Medchemexpress LLC Supplier No. HYN778110MM/1ML
(-)-(E)-Guggulsterone is an orally active natural stereoisomer that acts as an antagonist for the Farnesoid X Receptor (FXR). It exhibits potent hypolipidemic properties and suppresses dengue virus (DENV) replication by upregulating antiviral interferon responses. This compound also possesses antibacterial activities against Bacillus subtilis, Staphylococcus aureus, and Pseudomonas aeruginosa, and has cardiac protective and antioxidant activities.
  • Acts as an antagonist for Farnesoid X Receptor (FXR)
  • Exhibits potent hypolipidemic properties
  • Suppresses dengue virus (DENV) replication
  • Possesses antibacterial activities against Bacillus subtilis, Staphylococcus aureus, and Pseudomonas aeruginosa
  • Has cardiac protective and antioxidant activities