Learn More
E. HISTOLYTICA QUIK CHEK™
Catalog No. 23046790
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
23-046-790 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-046-790 Supplier Techlab Inc Supplier No. T30409
Add to Cart
Add to Cart
This item is not returnable. View return policy
The E. HISTOLYTICA QUIK CHEK™ test is a rapid test for the qualitative detection of Entamoeba histolytica in fecal samples and targets an adhesin unique to E. histolytica. The test can be used for quick and reliable diagnosis of amebiasis. Developed and Manufactured by TECHLAB, Inc.
  • Does not cross-react with E. dispar  detecting only pathogenic E. histolytica
  • Results in 30 minutes without instrumentation
  • More sensitive and specific than microscopy
  • Simple and clear interpretation of results
  • Specifications
    Detectable Analytes E. histolytica
    Type E. Histolytica QUIK CHEK™
    Includes 25 Membrane device pouches, diluent, wash buffer, conjugate, substrate, positive control, disposable plastic transfer pipettes
    Quantity 25 Tests