Learn More
Medchemexpress LLC (E)-KN-93 (phosphate) | 1188890-41-6 | MFCD03788201 | 99.9% | 599.03 g/mol | C26H32ClN2O8PS | 5 MG
SDP
Catalog No. 5000263852
Encompass_Preferred
(E)-KN-93 phosphate is a cell-permeable, competitive inhibitor of Ca2+/calmodulin-dependent protein kinase II (CaMKII). It disrupts Ca2+/CaM interaction with CaMKII and is supplied as a purified research reagent with analytical documentation for use in biochemical and cellular studies.
  • High purity (99.85%).
  • Water-soluble phosphate form for improved solubility.
  • Suitable for biochemical and cellular assays.
  • Provided with COA, HNMR, LCMS, and SDS documents.
  • Molecular formula C26H32ClN2O8PS; molecular weight 599.03 g/mol.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-63852 Supplier Medchemexpress LLC Supplier No. HY1101405MG
(E)-KN-93 phosphate is a cell-permeable, competitive inhibitor of Ca2+/calmodulin-dependent protein kinase II (CaMKII). It disrupts Ca2+/CaM interaction with CaMKII and is supplied as a purified research reagent with analytical documentation for use in biochemical and cellular studies.
  • High purity (99.85%).
  • Water-soluble phosphate form for improved solubility.
  • Suitable for biochemical and cellular assays.
  • Provided with COA, HNMR, LCMS, and SDS documents.
  • Molecular formula C26H32ClN2O8PS; molecular weight 599.03 g/mol.
  • Available in small research pack sizes for laboratory use.