Learn More
Medchemexpress LLC HY-112762A 10mg Medchemexpress, (E)-LHF-535 CAS: Purity:>98%
SDP
Catalog No. 501969164
Encompass_Preferred
Medchemexpress, HY-112762A 10mg (E)-LHF-535 CAS: (E)-LHF-535 is the E-isomer of LHF-535. LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38, has EC50s of <1 μM, <1 μM, <1 μM, and 1-10 μM for Lassa, Machupo, Junin, and VSVg virus, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9164 Supplier Medchemexpress LLC Supplier No. HY112762A10MG
Medchemexpress, HY-112762A 10mg (E)-LHF-535 CAS: (E)-LHF-535 is the E-isomer of LHF-535. LHF-535 is an antiviral agent extracted from patent WO2013123215A2, Compound 38, has EC50s of <1 μM, <1 μM, <1 μM, and 1-10 μM for Lassa, Machupo, Junin, and VSVg virus, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.