Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-LHF-535 | 2449213-57-2 | 99.2% | 412.52 | 1 ML
SDP

Catalog No. 5000437782
Encompass_Preferred

(E)-LHF-535 is the E-isomer of LHF-535, an antiviral agent. It demonstrates EC50s of less than 1 μM against Lassa, Machupo, and Junin viruses, and 1-10 μM against VSVg virus. This compound was extracted from patent WO2013123215A2.

  • E-isomer of LHF-535
  • Antiviral agent
  • Inhibits Lassa, Machupo, Junin viruses (EC50 <1 μM)
  • Inhibits VSVg virus (EC50 1-10 μM)
  • Molecular weight: 412.52
  • Formula: C27H28N2O2
  • Appearance: Solid, light yellow to yellow
  • Ships at room temperature (continental US)
  • Powder stable: 3 years at -20°C, 2 years at 4°C
  • Solution stable: 2 years at -80°C, 1 year at -20°C
  • Soluble in DMSO (60 mg/mL)
  • Research use for viral infections, particularly Arenavirus

Catalog No. 50-004-37782 Supplier Medchemexpress LLC Supplier No. HY11276210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(E)-LHF-535 is the E-isomer of LHF-535, an antiviral agent. It demonstrates EC50s of less than 1 μM against Lassa, Machupo, and Junin viruses, and 1-10 μM against VSVg virus. This compound was extracted from patent WO2013123215A2.

  • E-isomer of LHF-535
  • Antiviral agent
  • Inhibits Lassa, Machupo, Junin viruses (EC50 <1 μM)
  • Inhibits VSVg virus (EC50 1-10 μM)
  • Molecular weight: 412.52
  • Formula: C27H28N2O2
  • Appearance: Solid, light yellow to yellow
  • Ships at room temperature (continental US)
  • Powder stable: 3 years at -20°C, 2 years at 4°C
  • Solution stable: 2 years at -80°C, 1 year at -20°C
  • Soluble in DMSO (60 mg/mL)
  • Research use for viral infections, particularly Arenavirus

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.