Learn More
Medchemexpress LLC HY-N7127 100mg Medchemexpress, (E)-m-Coumaric acid CAS:14755-02-3 Purity:>98%
SDP
Catalog No. 501970508
Encompass_Preferred
Medchemexpress, HY-N7127 100mg (E)-m-Coumaric acid CAS:14755-02-3 (E)-m-Coumaric acid (3-Hydroxycinnamic acid) is an aromatic acid that highly abundant in food. (E)-m-Coumaric acid (3-Hydroxycinnamic acid) is an antioxidant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0508 Supplier Medchemexpress LLC Supplier No. HYN7127100MG
Medchemexpress, HY-N7127 100mg (E)-m-Coumaric acid CAS:14755-02-3 (E)-m-Coumaric acid (3-Hydroxycinnamic acid) is an aromatic acid that highly abundant in food. (E)-m-Coumaric acid (3-Hydroxycinnamic acid) is an antioxidant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.