Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-m-Coumaric acid | 14755-02-3 | 1 ML
SDP

Catalog No. 5000418131
Encompass_Preferred

(E)-m-Coumaric acid (3-Hydroxycinnamic acid) is an aromatic acid highly abundant in food that acts as an antioxidant. It is categorized as an endogenous and microbial metabolite, targeting metabolic enzyme/protease pathways, and is intended for research use only.

  • Aromatic acid highly abundant in food
  • Acts as an antioxidant
  • Categorized as an endogenous metabolite
  • Categorized as a microbial metabolite
  • Targets metabolic enzyme/protease pathways
  • Purity of 99.74%
  • Appearance as an off-white to yellow solid
  • Molecular formula C9H8O3
  • Molecular weight 164.16
  • Solubility in DMSO: 100 mg/mL
  • Recommended storage at 4°C, protected from light

Catalog No. 50-004-18131 Supplier Medchemexpress LLC Supplier No. HYN712710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-m-Coumaric acid (3-Hydroxycinnamic acid) is an aromatic acid highly abundant in food that acts as an antioxidant. It is categorized as an endogenous and microbial metabolite, targeting metabolic enzyme/protease pathways, and is intended for research use only.

  • Aromatic acid highly abundant in food
  • Acts as an antioxidant
  • Categorized as an endogenous metabolite
  • Categorized as a microbial metabolite
  • Targets metabolic enzyme/protease pathways
  • Purity of 99.74%
  • Appearance as an off-white to yellow solid
  • Molecular formula C9H8O3
  • Molecular weight 164.16
  • Solubility in DMSO: 100 mg/mL
  • Recommended storage at 4°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.