Learn More
Medchemexpress LLC (E)-Methyl 4-coumarate | 19367-38-5 | 25 G
SDP
Catalog No. 5000421926
Encompass_Preferred
E-Methyl 4-coumarate is a phenolic compound and a derivative of Cinnamic acid, found in plants such as *Allium cepa* and *Morinda citrifolia L.*. It inhibits GSK3β activity and modulates inflammatory cytokine levels by increasing IL-10 and decreasing IL-4. When combined with Carnosic acid, it induces apoptosis and exhibits anticancer effects against acute myeloid leukemia. Additionally, it ameliorates *Plasmodium berghei* NK65 infection.
  • Inhibits GSK3β activity
  • Modulates inflammatory cytokine levels
  • Increases IL-10 and decreases IL-4
  • Induces apoptosis when combined with carnosic acid
  • Exhibits anticancer effects against acute myeloid leukemia with carnosic acid
  • Ameliorates *Plasmodium berghei* NK65 infection
Catalog No. 50-004-21926 Supplier Medchemexpress LLC Supplier No. 25GHYN2492
E-Methyl 4-coumarate is a phenolic compound and a derivative of Cinnamic acid, found in plants such as *Allium cepa* and *Morinda citrifolia L.*. It inhibits GSK3β activity and modulates inflammatory cytokine levels by increasing IL-10 and decreasing IL-4. When combined with Carnosic acid, it induces apoptosis and exhibits anticancer effects against acute myeloid leukemia. Additionally, it ameliorates *Plasmodium berghei* NK65 infection.
  • Inhibits GSK3β activity
  • Modulates inflammatory cytokine levels
  • Increases IL-10 and decreases IL-4
  • Induces apoptosis when combined with carnosic acid
  • Exhibits anticancer effects against acute myeloid leukemia with carnosic acid
  • Ameliorates *Plasmodium berghei* NK65 infection