Learn More
Medchemexpress LLC (E)-N-(3-methyl-1,1-dioxo-1,4-thiazinan-4-yl)-1-(5-nitrofuran-2-yl)methanimine | 23256-30-6 | MFCD00869254 | 99.7% | 287.29 g/mol | C10H13N3O5S | 100 MG
SDP
Catalog No. 5000301239
Encompass_Preferred
Nifurtimox is a nitrofuran antiprotozoal compound provided as a research-grade chemical for laboratory studies. It is commonly used in experimental models of Trypanosoma cruzi infection and in preclinical studies such as neuroblastoma research and enzyme activity assays.
  • Antiprotozoal nitrofuran compound for research applications.
  • Used in Trypanosoma cruzi infection models and neuroblastoma studies.
  • Reported to modulate lactate dehydrogenase (LDH) activity in assays.
  • Reported purity 99.72%.
  • Molecular formula C10H13N3O5S and molar mass ~287.29 g/mol.
  • Available as a 100 mg research pack.
Catalog No. 50-003-01239 Supplier Medchemexpress LLC Supplier No. HYW040073100MG
Nifurtimox is a nitrofuran antiprotozoal compound provided as a research-grade chemical for laboratory studies. It is commonly used in experimental models of Trypanosoma cruzi infection and in preclinical studies such as neuroblastoma research and enzyme activity assays.
  • Antiprotozoal nitrofuran compound for research applications.
  • Used in Trypanosoma cruzi infection models and neuroblastoma studies.
  • Reported to modulate lactate dehydrogenase (LDH) activity in assays.
  • Reported purity 99.72%.
  • Molecular formula C10H13N3O5S and molar mass ~287.29 g/mol.
  • Available as a 100 mg research pack.