Learn More
Medchemexpress LLC (E)-N-(4-(1-(3-Aminobenzoyl)piperidin-4-yl)butyl)-3-(pyridin-3-yl)acrylamide | 2222981-61-3 | 99.6% | 406.52 | 50 MG
SDP
Catalog No. 5000405147
Encompass_Preferred
Nampt-IN-15 is an Nampt inhibitor. It demonstrates cytotoxicity against BxPC-3, HepG2, L540cy, and MOLM-13 cell lines, with IC50 values of 38.5 nM, 8 nM, 8.5 nM, and 7 nM respectively. It is intended for research use only.
  • Nampt inhibitor.
  • Shows cytotoxicity against various cell lines: BxPC-3 (IC50: 38.5 nM), HepG2 (IC50: 8 nM), L540cy (IC50: 8.5 nM), and MOLM-13 (IC50: 7 nM).
Catalog No. 50-004-05147 Supplier Medchemexpress LLC Supplier No. HY40223750MG
Nampt-IN-15 is an Nampt inhibitor. It demonstrates cytotoxicity against BxPC-3, HepG2, L540cy, and MOLM-13 cell lines, with IC50 values of 38.5 nM, 8 nM, 8.5 nM, and 7 nM respectively. It is intended for research use only.
  • Nampt inhibitor.
  • Shows cytotoxicity against various cell lines: BxPC-3 (IC50: 38.5 nM), HepG2 (IC50: 8 nM), L540cy (IC50: 8.5 nM), and MOLM-13 (IC50: 7 nM).