Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-N-(5-bromo-2-methoxyphenyl)sulfonyl-3-[5-chloro-2-(naphthalen-2-ylmethyl)phenyl]prop-2-enamide | 244101-03-9 | 98.0% | 570.88 g/mol | C27H21BrClNO4S | 10 MG
SDP

Catalog No. 5000298364
Encompass_Preferred

L-826266 is a selective and competitive EP3 (prostaglandin E receptor 3) antagonist for research use. It has been applied in in vitro hippocampal slice studies and in vivo rodent seizure models to investigate prostaglandin signaling and convulsive disorders.

  • Selective EP3 receptor antagonist activity
  • Shown to delay seizures and increase seizure latency in PTZ rodent models
  • Prevents PGE2-induced decrease of Na+,K+-ATPase activity in hippocampal slices
  • High purity (98.0%) suitable for research applications
  • Molecular weight 570.88 g/mol; formula C27H21BrClNO4S
  • CAS number 244101-03-9 for unambiguous identification

Catalog No. 50-002-98364 Supplier Medchemexpress LLC Supplier No. HY1936110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-826266 is a selective and competitive EP3 (prostaglandin E receptor 3) antagonist for research use. It has been applied in in vitro hippocampal slice studies and in vivo rodent seizure models to investigate prostaglandin signaling and convulsive disorders.

  • Selective EP3 receptor antagonist activity
  • Shown to delay seizures and increase seizure latency in PTZ rodent models
  • Prevents PGE2-induced decrease of Na+,K+-ATPase activity in hippocampal slices
  • High purity (98.0%) suitable for research applications
  • Molecular weight 570.88 g/mol; formula C27H21BrClNO4S
  • CAS number 244101-03-9 for unambiguous identification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.