Learn More
Medchemexpress LLC E-N-ethyl-N-(6,6-dimethyl-2-hepten-4-ynyl)-3-(2-methyl-2-(3-thienylmethoxy)propyloxy)benzylamine HCl | 204067-52-7 | 98.8% | 476.11 | C27H38ClNO2S | 100 MG
SDP
Catalog No. 5000214345
Encompass_Preferred
FR194738 is a research compound that potently inhibits squalene epoxidase and is used to investigate cholesterol biosynthesis in biochemical and cell-based assays. Reported activity includes an IC50 of 9.8 nM in HepG2 cell homogenates and 4.9 nM for inhibition of cholesterol synthesis in intact HepG2 cells. Supplied as a high-purity free base with multiple package sizes and storage recommendations for laboratory use.
  • Potent squalene epoxidase inhibition in biochemical and cell assays.
  • Demonstrated low-nanomolar IC50 in HepG2 assays.
  • Available as a free base with supplier-reported high purity.
  • Multiple package sizes for flexible experimental scaling.
  • Storage recommendations support compound stability for routine use.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-14345 Supplier Medchemexpress LLC Supplier No. HY100303A100MG
FR194738 is a research compound that potently inhibits squalene epoxidase and is used to investigate cholesterol biosynthesis in biochemical and cell-based assays. Reported activity includes an IC50 of 9.8 nM in HepG2 cell homogenates and 4.9 nM for inhibition of cholesterol synthesis in intact HepG2 cells. Supplied as a high-purity free base with multiple package sizes and storage recommendations for laboratory use.
  • Potent squalene epoxidase inhibition in biochemical and cell assays.
  • Demonstrated low-nanomolar IC50 in HepG2 assays.
  • Available as a free base with supplier-reported high purity.
  • Multiple package sizes for flexible experimental scaling.
  • Storage recommendations support compound stability for routine use.
  • For research use only; not for human or clinical use.