Learn More
TARGETMOL CHEMICALS INC Research Chemical 10mg
SDP
Catalog No. 503038160
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Naringenin chalcone (Isosalipurpol) has antiallergic activity Naringenin chalcone suppresses asthmatic symptoms by inhibiting Th2 cytokine production from CD4 T cells. Naringenin chalcone is a potent tomato flavonoid that improves adipocyte metabolic functions and exerts insulin-sensitizing effects by activating an adiponectin-related pathway. Naringenin chalcone exhibits anti-inflammatory properties by inhibiting the production of proinflammatory cytokines in the interaction between adipocytes and macrophages, may be useful for ameliorating the inflammatory changes in obese adipose tissue. purity: 99%
Catalog No. 50-303-8160 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S217310MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Naringenin chalcone (Isosalipurpol) has antiallergic activity Naringenin chalcone suppresses asthmatic symptoms by inhibiting Th2 cytokine production from CD4 T cells. Naringenin chalcone is a potent tomato flavonoid that improves adipocyte metabolic functions and exerts insulin-sensitizing effects by activating an adiponectin-related pathway. Naringenin chalcone exhibits anti-inflammatory properties by inhibiting the production of proinflammatory cytokines in the interaction between adipocytes and macrophages, may be useful for ameliorating the inflammatory changes in obese adipose tissue. purity: 99%