Learn More
Medchemexpress LLC (E)-Naringenin chalcone | 25515-46-2 | 100 MG
SDP
Catalog No. 5000421734
Encompass_Preferred
(E)-Naringenin chalcone is an orally active anti-allergic agent that also possesses antioxidant and anti-inflammatory activities. It can improve adipocyte functions and inhibits histamine release from rat peritoneal mast cells.
  • Inhibits histamine release from rat peritoneal mast cells.
  • Improves adipocyte functions by enhancing adiponectin production.
  • Stimulates the activity of PPARγ in cells.
  • Inhibits the production of TNF-alpha, MCP-1, and nitric oxide (NO) by LPS-stimulated macrophages.
  • Exhibits anti-allergic effects in type I allergic mice.
  • Suppresses allergic asthma by inhibiting the type-2 function of CD4 T cells in allergic airway inflammatory mice.
Catalog No. 50-004-21734 Supplier Medchemexpress LLC Supplier No. HYN3007A100MG
(E)-Naringenin chalcone is an orally active anti-allergic agent that also possesses antioxidant and anti-inflammatory activities. It can improve adipocyte functions and inhibits histamine release from rat peritoneal mast cells.
  • Inhibits histamine release from rat peritoneal mast cells.
  • Improves adipocyte functions by enhancing adiponectin production.
  • Stimulates the activity of PPARγ in cells.
  • Inhibits the production of TNF-alpha, MCP-1, and nitric oxide (NO) by LPS-stimulated macrophages.
  • Exhibits anti-allergic effects in type I allergic mice.
  • Suppresses allergic asthma by inhibiting the type-2 function of CD4 T cells in allergic airway inflammatory mice.