Learn More
Medchemexpress LLC (E)-Naringenin chalcone | 25515-46-2 | 25 MG
SDP
Catalog No. 5000421587
Encompass_Preferred
(E)-Naringenin chalcone is an orally active anti-allergic agent that exhibits antioxidant and anti-inflammatory properties. It can enhance adipocyte functions and inhibits the release of histamine from rat peritoneal mast cells.
  • Anti-allergic agent: orally active and inhibits histamine release from rat peritoneal mast cells.
  • Antioxidant activity: possesses antioxidant properties.
  • Anti-inflammatory activity: demonstrates anti-inflammatory effects.
  • Adipocyte function improvement: can improve the functions of adipocytes.
  • PPARγ stimulation: stimulates the activity of PPARγ in NIH-3T3 cells.
  • Inhibition of inflammatory mediators: inhibits the production of TNF-alpha, MCP-1, and nitric oxide (NO) by LPS-stimulated RAW 264 macrophages.
  • Suppresses allergic asthma: inhibits the type-2 function of CD4 T cells in allergic airway inflammatory mice.
Catalog No. 50-004-21587 Supplier Medchemexpress LLC Supplier No. 25MGHYN3007A
(E)-Naringenin chalcone is an orally active anti-allergic agent that exhibits antioxidant and anti-inflammatory properties. It can enhance adipocyte functions and inhibits the release of histamine from rat peritoneal mast cells.
  • Anti-allergic agent: orally active and inhibits histamine release from rat peritoneal mast cells.
  • Antioxidant activity: possesses antioxidant properties.
  • Anti-inflammatory activity: demonstrates anti-inflammatory effects.
  • Adipocyte function improvement: can improve the functions of adipocytes.
  • PPARγ stimulation: stimulates the activity of PPARγ in NIH-3T3 cells.
  • Inhibition of inflammatory mediators: inhibits the production of TNF-alpha, MCP-1, and nitric oxide (NO) by LPS-stimulated RAW 264 macrophages.
  • Suppresses allergic asthma: inhibits the type-2 function of CD4 T cells in allergic airway inflammatory mice.