Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-Naringenin chalcone | 25515-46-2 | 5 MG
SDP

Catalog No. 5000421315
Encompass_Preferred

(E)-Naringenin chalcone is an orally active anti-allergic agent. It also possesses antioxidant and anti-inflammatory activities, and can improve adipocyte functions. It inhibits histamine release from rat peritoneal mast cells.

  • Anti-allergic agent: Inhibits histamine release from mast cells and reduces ear-swelling in allergic mice.
  • Antioxidant activity: Demonstrated antioxidant properties.
  • Anti-inflammatory activity: Inhibits production of TNF-alpha, MCP-1, and nitric oxide by LPS-stimulated macrophages.
  • Adipocyte function improvement: Enhances adiponectin production and stimulates PPARγ activity.
  • High purity: 98.87%.
  • Appearance: Solid, light yellow to yellow.
  • Molecular weight: 272.26.
  • Chemical formula: C15H12O5.
  • Initial source: Plants (Solanaceae, *Solanum lycopersicum L.*).

Catalog No. 50-004-21315 Supplier Medchemexpress LLC Supplier No. HYN3007A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-Naringenin chalcone is an orally active anti-allergic agent. It also possesses antioxidant and anti-inflammatory activities, and can improve adipocyte functions. It inhibits histamine release from rat peritoneal mast cells.

  • Anti-allergic agent: Inhibits histamine release from mast cells and reduces ear-swelling in allergic mice.
  • Antioxidant activity: Demonstrated antioxidant properties.
  • Anti-inflammatory activity: Inhibits production of TNF-alpha, MCP-1, and nitric oxide by LPS-stimulated macrophages.
  • Adipocyte function improvement: Enhances adiponectin production and stimulates PPARγ activity.
  • High purity: 98.87%.
  • Appearance: Solid, light yellow to yellow.
  • Molecular weight: 272.26.
  • Chemical formula: C15H12O5.
  • Initial source: Plants (Solanaceae, *Solanum lycopersicum L.*).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.