Learn More
Medchemexpress LLC (E)-naringenin chalcone | 25515-46-2 | MFCD00016762 | 98.9% | 272.26 g/mol | C15H12O5 | 10 MG
SDP
Catalog No. 5000289392
Encompass_Preferred
(E)-Naringenin chalcone is a flavonoid-derived chalcone supplied for research use. It is described as an orally active anti-allergic agent with reported antioxidant, anti-inflammatory, and anticancer activities. The compound is provided as a light yellow solid with high purity and is intended for laboratory studies and small-molecule assays.
  • Orally active anti-allergic agent and antioxidant.
  • Reported anti-inflammatory and anticancer activity for research studies.
  • Light yellow solid appearance suitable for handling in laboratory assays.
  • High purity (~98.9%) for reproducible experimental results.
  • Molecular weight 272.26 g/mol; formula C15H12O5.
  • Store sealed away from moisture and light; recommended cold storage conditions for solutions.
Catalog No. 50-002-89392 Supplier Medchemexpress LLC Supplier No. HYN3007A10MG
(E)-Naringenin chalcone is a flavonoid-derived chalcone supplied for research use. It is described as an orally active anti-allergic agent with reported antioxidant, anti-inflammatory, and anticancer activities. The compound is provided as a light yellow solid with high purity and is intended for laboratory studies and small-molecule assays.
  • Orally active anti-allergic agent and antioxidant.
  • Reported anti-inflammatory and anticancer activity for research studies.
  • Light yellow solid appearance suitable for handling in laboratory assays.
  • High purity (~98.9%) for reproducible experimental results.
  • Molecular weight 272.26 g/mol; formula C15H12O5.
  • Store sealed away from moisture and light; recommended cold storage conditions for solutions.