Learn More
Medchemexpress LLC HY-100573 5mg Medchemexpress, (E)-Necrosulfonamide CAS:1360614-48-7 Purity:>98%
SDP
Catalog No. 501931886
Encompass_Preferred
Medchemexpress, HY-100573 5mg (E)-Necrosulfonamide CAS:1360614-48-7 (E)-Necrosulfonamide is a necroptosis inhibitor acting by selectively targeting the mixed lineage kinase domain-like protein (MLKL) to block the necrosome formation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1886 Supplier Medchemexpress LLC Supplier No. HY1005735MG
Medchemexpress, HY-100573 5mg (E)-Necrosulfonamide CAS:1360614-48-7 (E)-Necrosulfonamide is a necroptosis inhibitor acting by selectively targeting the mixed lineage kinase domain-like protein (MLKL) to block the necrosome formation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.