Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-oct-2-enoic acid | 1871-67-6 | MFCD00002706 | 98.3% | 142.20 g/mol | C8H14O2 | 1 ML
SDP

Catalog No. 5000210160
Encompass_Preferred

(E)-Oct-2-enoic acid is an endogenous metabolite available as a 10 mM solution in DMSO or as solid quantities for research use. It has CAS number 1871-67-6, molecular formula C8H14O2, molecular weight 142.20 g/mol, and reported purity of 98.3%.

  • Supplied as 10 mM, 1 mL solution in DMSO and as 100 mg, 500 mg, 1 g, and 5 g solids.
  • Purity 98.3%.
  • Solubility in DMSO 100 mg/mL; ultrasonic assistance recommended and use newly opened DMSO.
  • Physical state liquid; density approximately 0.944 g/cm3.
  • Includes downloadable data sheet, certificate of analysis, safety data sheet, HNMR, and LCMS files.

Catalog No. 50-002-10160 Supplier Medchemexpress LLC Supplier No. HYW04690610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-Oct-2-enoic acid is an endogenous metabolite available as a 10 mM solution in DMSO or as solid quantities for research use. It has CAS number 1871-67-6, molecular formula C8H14O2, molecular weight 142.20 g/mol, and reported purity of 98.3%.

  • Supplied as 10 mM, 1 mL solution in DMSO and as 100 mg, 500 mg, 1 g, and 5 g solids.
  • Purity 98.3%.
  • Solubility in DMSO 100 mg/mL; ultrasonic assistance recommended and use newly opened DMSO.
  • Physical state liquid; density approximately 0.944 g/cm3.
  • Includes downloadable data sheet, certificate of analysis, safety data sheet, HNMR, and LCMS files.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.