Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-osmundacetone | 123694-03-1 | 99.9% | 178.19 g/mol | C10H10O3 | 10 MG
SDP

Catalog No. 5000296334
Encompass_Preferred

(E)-Osmundacetone is the E-isomer of osmundacetone, a small-molecule research compound reported to suppress phosphorylation of MAPK family kinases including p38, JNK, and ERK. It is used in biochemical and cell-based assays to study MAPK/ERK signaling and oxidative stress responses; intended for laboratory research use only.

  • CAS number 123694-03-1.
  • Molecular formula C10H10O3.
  • Molecular weight 178.19 g/mol.
  • High purity 99.9%.
  • Reported activity: suppresses phosphorylation of p38, JNK, and ERK.
  • Suitable for in vitro biochemical and cell-based assays.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-96334 Supplier Medchemexpress LLC Supplier No. HYN196610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-Osmundacetone is the E-isomer of osmundacetone, a small-molecule research compound reported to suppress phosphorylation of MAPK family kinases including p38, JNK, and ERK. It is used in biochemical and cell-based assays to study MAPK/ERK signaling and oxidative stress responses; intended for laboratory research use only.

  • CAS number 123694-03-1.
  • Molecular formula C10H10O3.
  • Molecular weight 178.19 g/mol.
  • High purity 99.9%.
  • Reported activity: suppresses phosphorylation of p38, JNK, and ERK.
  • Suitable for in vitro biochemical and cell-based assays.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.