Learn More
Medchemexpress LLC (E)-p-Coumaramide | 194940-15-3 | 163.17 | 1 MG
SDP
Catalog No. 5000416182
Encompass_Preferred
(E)-p-Coumaramide, isolated from Welsh onion, acts as an α-Glucosidase inhibitor. It has a molecular weight of 163.17 and a chemical formula of C9H9NO2. The substance appears as a light yellow to yellow solid.
  • Functions as an α-Glucosidase inhibitor.
  • Intended for research use only.
  • Not sold to patients.
Catalog No. 50-004-16182 Supplier Medchemexpress LLC Supplier No. HYN19011MG
(E)-p-Coumaramide, isolated from Welsh onion, acts as an α-Glucosidase inhibitor. It has a molecular weight of 163.17 and a chemical formula of C9H9NO2. The substance appears as a light yellow to yellow solid.
  • Functions as an α-Glucosidase inhibitor.
  • Intended for research use only.
  • Not sold to patients.