Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E)-p-Coumaramide | 194940-15-3 | 163.17 | 5 MG
SDP

Catalog No. 5000415073
Encompass_Preferred

(E)-p-Coumaramide is an α-Glucosidase inhibitor that can be isolated from Welsh onion. It is for research use only and not sold to patients.

  • Alpha-glucosidase inhibitor
  • Isolated from Welsh onion
  • Target is glycosidase
  • Pathway is metabolic enzyme/protease
  • Purity of 95.0%
  • Appearance as a solid
  • Color is light yellow to yellow

Catalog No. 50-004-15073 Supplier Medchemexpress LLC Supplier No. HYN19015MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E)-p-Coumaramide is an α-Glucosidase inhibitor that can be isolated from Welsh onion. It is for research use only and not sold to patients.

  • Alpha-glucosidase inhibitor
  • Isolated from Welsh onion
  • Target is glycosidase
  • Pathway is metabolic enzyme/protease
  • Purity of 95.0%
  • Appearance as a solid
  • Color is light yellow to yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.