Learn More
Agilent Technologies™ E-Plate VIEW 16
Description
- Derive cell kinetics
- Obtain continuous immune cell killing assay time points during the entire experiment
- RTCA provides a continuous quantitative readout of proliferation rate, cell number, size/shape, and substrate attachment quality
Specifications
Specifications
| Barcode | Includes Barcode |
| Format | Standard |
| Array | 2 x 8 |
| Color | Clear, Gold |
| Disposable | Single-use |
| Material | Polystyrene |
| Volume (Metric) Well | 270 ±10 μL |
| No. of Wells | 16 |
| Sterility | Non-sterile |
| Surface Treatment | Gold-plated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.