Learn More
MSC E.R. Wagner Caster, 5 in Wheel Dia, Soft Rubber, Swivel, Threaded Stem
Catalog No. 5000513518
Encompass
This caster is designed with a 5 in diameter, 1-1/4 in wide wheel, and a 5-1/2 in overall height stem mount. It features soft rubber, a 190 lb capacity, Delrin bearing, and a 1/2-13 x 1 in threaded stem.
  • Threaded stem type
  • 1/2-13 stem diameter
  • Swivel design
  • Soft rubber wheel material
  • 5 in wheel diameter
  • Hardened-heat treated construction
  • Zinc chromate finish
  • Delrin bearing type
  • Soft wheel hardness
  • Suitable for industrial and commercial applications
  • 190 lb capacity
  • 1/2-13 x 1 in threaded stem
Catalog No. 50-005-13518 Supplier MSC Supplier No. 01505825
This caster is designed with a 5 in diameter, 1-1/4 in wide wheel, and a 5-1/2 in overall height stem mount. It features soft rubber, a 190 lb capacity, Delrin bearing, and a 1/2-13 x 1 in threaded stem.
  • Threaded stem type
  • 1/2-13 stem diameter
  • Swivel design
  • Soft rubber wheel material
  • 5 in wheel diameter
  • Hardened-heat treated construction
  • Zinc chromate finish
  • Delrin bearing type
  • Soft wheel hardness
  • Suitable for industrial and commercial applications
  • 190 lb capacity
  • 1/2-13 x 1 in threaded stem