Learn More
MSC E.R. Wagner Caster, Stem Mount Swivel, 3-1/2 in Polyolefin, 250 Lb Capacity
Catalog No. 5000533247
Encompass
This swivel caster features a 3-1/2 inch diameter polyolefin wheel and a 1/2-13 x 1-1/2 inch threaded stem, designed for smooth movement on carpeted surfaces. With a 250 pound load capacity and Delrin bearings, it offers reliable performance for various applications.
  • Swivel design for easy maneuverability
  • Delrin bearing for smooth operation
  • 1/2-13 x 1-1/2 inch threaded stem for secure mounting
  • Hardened and heat-treated construction for durability
  • Zinc finish for corrosion resistance
  • NSF certified for quality and safety
  • Ideal for use on carpeted surfaces
  • 3-1/2 inch diameter polyolefin wheel
  • 250 pound load capacity
Catalog No. 50-005-33247 Supplier MSC Supplier No. 01506682
This swivel caster features a 3-1/2 inch diameter polyolefin wheel and a 1/2-13 x 1-1/2 inch threaded stem, designed for smooth movement on carpeted surfaces. With a 250 pound load capacity and Delrin bearings, it offers reliable performance for various applications.
  • Swivel design for easy maneuverability
  • Delrin bearing for smooth operation
  • 1/2-13 x 1-1/2 inch threaded stem for secure mounting
  • Hardened and heat-treated construction for durability
  • Zinc finish for corrosion resistance
  • NSF certified for quality and safety
  • Ideal for use on carpeted surfaces
  • 3-1/2 inch diameter polyolefin wheel
  • 250 pound load capacity