Learn More
MSC E.R. Wagner Caster, Stem Mount Swivel, 3" Wheel, Polyurethane, 210 Lb Capacity
Catalog No. 5000534158
Encompass
This E.R. Wagner stem mount swivel caster is designed for industrial and commercial applications, providing reliable mobility with a 210 Lb capacity.
  • Threaded stem for secure mounting
  • Swivel design for easy maneuverability
  • 3 inch wheel diameter
  • Polyurethane and rubber wheel material for durability
  • Delrin bearing type ensures smooth operation
  • Hardened-heat treated construction for strength
  • Zinc chromate finish for corrosion resistance
  • 1/2-13 x 1 inch threaded stem dimensions
Catalog No. 50-005-34158 Supplier MSC Supplier No. 01506054
This E.R. Wagner stem mount swivel caster is designed for industrial and commercial applications, providing reliable mobility with a 210 Lb capacity.
  • Threaded stem for secure mounting
  • Swivel design for easy maneuverability
  • 3 inch wheel diameter
  • Polyurethane and rubber wheel material for durability
  • Delrin bearing type ensures smooth operation
  • Hardened-heat treated construction for strength
  • Zinc chromate finish for corrosion resistance
  • 1/2-13 x 1 inch threaded stem dimensions