Learn More
MSC E.R. Wagner Rigid Caster, Polyolefin, 4 inch Wheel, 375 lb Capacity
Catalog No. 5000529824
Encompass
This rigid top plate caster features a polyolefin wheel, designed for various applications including carpeted surfaces. It offers durability and reliable performance with a hardened-heat treated construction and a nickel finish.
  • Hardened-heat treated construction
  • Nickel finish
  • Suitable for carpeted surfaces
  • Delrin bearing
  • 4 inch wheel diameter
  • 1-3/8 inch wheel width
  • 5-3/8 inch overall height
  • 375 lb capacity
  • 3-1/8 x 4-1/8 inch plate size
Catalog No. 50-005-29824 Supplier MSC Supplier No. 01507813
This rigid top plate caster features a polyolefin wheel, designed for various applications including carpeted surfaces. It offers durability and reliable performance with a hardened-heat treated construction and a nickel finish.
  • Hardened-heat treated construction
  • Nickel finish
  • Suitable for carpeted surfaces
  • Delrin bearing
  • 4 inch wheel diameter
  • 1-3/8 inch wheel width
  • 5-3/8 inch overall height
  • 375 lb capacity
  • 3-1/8 x 4-1/8 inch plate size