Learn More
MSC E.R. Wagner Rigid Top Plate Caster, Hard Rubber, 210 lb Capacity
Catalog No. 5000533568
Encompass
This rigid top plate caster is designed for various material handling applications. It features a durable hard rubber wheel (3 in. diameter, 1-1/4 in. width) and offers a load capacity of 210 lb. The caster includes hardened-heat treated construction and a zinc finish for corrosion resistance, ensuring stability and controlled movement in industrial and commercial environments, including food service.
  • Rigid style ensures directional stability.
  • Hard rubber wheel provides durability and smooth operation.
  • 3 in. wheel diameter for compact design.
  • 1-1/4 in. wheel width for balanced load distribution.
  • 210 lb load capacity supports heavy-duty applications.
  • Delrin bearing type offers reliable performance.
  • Hardened-heat treated construction for extended service life.
  • Zinc finish/coating provides corrosion resistance.
  • Suitable for food service applications.
Catalog No. 50-005-33568 Supplier MSC Supplier No. 01507904
This rigid top plate caster is designed for various material handling applications. It features a durable hard rubber wheel (3 in. diameter, 1-1/4 in. width) and offers a load capacity of 210 lb. The caster includes hardened-heat treated construction and a zinc finish for corrosion resistance, ensuring stability and controlled movement in industrial and commercial environments, including food service.
  • Rigid style ensures directional stability.
  • Hard rubber wheel provides durability and smooth operation.
  • 3 in. wheel diameter for compact design.
  • 1-1/4 in. wheel width for balanced load distribution.
  • 210 lb load capacity supports heavy-duty applications.
  • Delrin bearing type offers reliable performance.
  • Hardened-heat treated construction for extended service life.
  • Zinc finish/coating provides corrosion resistance.
  • Suitable for food service applications.