Learn More
MSC E.R. Wagner Rigid Top Plate Caster, Phenolic, 6 in, 700 lb
Catalog No. 5000535297
Encompass
This rigid top plate caster features a durable phenolic wheel, making it suitable for a variety of industrial and institutional applications. With a 6-inch wheel diameter and a robust 700 lb load capacity, it provides reliable mobility for equipment and carts.
  • Phenolic wheel material for durability.
  • 6 inch wheel diameter and 2 inch wheel width.
  • 700 lb load capacity for heavy-duty use.
  • Overall height of 7-1/2 inches.
  • Top plate mount type for secure installation.
  • Equipped with a thread guard.
  • Roller bearing type ensures smooth operation.
  • Stamp formed construction with chrome zinc finish.
Catalog No. 50-005-35297 Supplier MSC Supplier No. 57078669
This rigid top plate caster features a durable phenolic wheel, making it suitable for a variety of industrial and institutional applications. With a 6-inch wheel diameter and a robust 700 lb load capacity, it provides reliable mobility for equipment and carts.
  • Phenolic wheel material for durability.
  • 6 inch wheel diameter and 2 inch wheel width.
  • 700 lb load capacity for heavy-duty use.
  • Overall height of 7-1/2 inches.
  • Top plate mount type for secure installation.
  • Equipped with a thread guard.
  • Roller bearing type ensures smooth operation.
  • Stamp formed construction with chrome zinc finish.