Learn More
MSC E.R. Wagner Rigid Top Plate Caster, Polyolefin, 4 in Wheel Dia, 1-3/8 in Wheel Width, 300 lb Capacity
Catalog No. 5000533506
Encompass
This Rigid Top Plate Caster features a polyolefin wheel with a 4 inch diameter and 1-3/8 inch width. It offers a load capacity of 300 lb and an overall height of 5-1/16 inches.
  • Plate mount type
  • 4 in wheel diameter
  • Polyolefin wheel material
  • 1-3/8 in wheel width
  • Black wheel color
  • Roller bearing type
  • Hardened-heat treated construction
  • 4.625 in plate length
  • 3.75 in plate width
  • 5-1/16 in overall height
  • Zinc finish
  • 300 lb load capacity
  • Suitable for carpeted surfaces
  • Rigid style
Catalog No. 50-005-33506 Supplier MSC Supplier No. 01507490
This Rigid Top Plate Caster features a polyolefin wheel with a 4 inch diameter and 1-3/8 inch width. It offers a load capacity of 300 lb and an overall height of 5-1/16 inches.
  • Plate mount type
  • 4 in wheel diameter
  • Polyolefin wheel material
  • 1-3/8 in wheel width
  • Black wheel color
  • Roller bearing type
  • Hardened-heat treated construction
  • 4.625 in plate length
  • 3.75 in plate width
  • 5-1/16 in overall height
  • Zinc finish
  • 300 lb load capacity
  • Suitable for carpeted surfaces
  • Rigid style