Learn More
MSC E.R. Wagner Rigid Top Plate Caster, Thermoplastic Rubber, 3 in Wheel, 125 lb Capacity
Catalog No. 5000529586
Encompass
This rigid top plate caster features a 3 inch diameter thermoplastic rubber wheel with a 1-1/4 inch width, offering a load capacity of 125 lb. Designed for hard surfaces, it has an overall height of 3-3/4 inches and a zinc finish. Its hardened-heat treated construction ensures durability.
  • Top plate mount
  • 3 inch wheel diameter
  • Thermoplastic rubber wheel material
  • 1-1/4 inch wheel width
  • Gray wheel color
  • Delrin bearing type
  • Hardened-heat treated construction
  • Zinc finish
  • 125 lb load capacity
  • Corrosion-resistant
  • Suitable for hard surfaces
  • Rigid style
Catalog No. 50-005-29586 Supplier MSC Supplier No. 01507375
This rigid top plate caster features a 3 inch diameter thermoplastic rubber wheel with a 1-1/4 inch width, offering a load capacity of 125 lb. Designed for hard surfaces, it has an overall height of 3-3/4 inches and a zinc finish. Its hardened-heat treated construction ensures durability.
  • Top plate mount
  • 3 inch wheel diameter
  • Thermoplastic rubber wheel material
  • 1-1/4 inch wheel width
  • Gray wheel color
  • Delrin bearing type
  • Hardened-heat treated construction
  • Zinc finish
  • 125 lb load capacity
  • Corrosion-resistant
  • Suitable for hard surfaces
  • Rigid style