Learn More
MSC E.R. Wagner Stem Mount Swivel Caster, 2 in. Hard Rubber, 100 Lb
Catalog No. 5000534163
Encompass
This E.R. Wagner stem mount swivel caster features a 2 in diameter hard rubber wheel and a chrome finish, designed for various applications with a 100 Lb capacity.
  • Hard rubber wheel
  • 100 Lb capacity
  • Delrin bearing
  • 5/16-18 x 1 in threaded stem
  • Hassle free guarantee
Catalog No. 50-005-34163 Supplier MSC Supplier No. 01505015
This E.R. Wagner stem mount swivel caster features a 2 in diameter hard rubber wheel and a chrome finish, designed for various applications with a 100 Lb capacity.
  • Hard rubber wheel
  • 100 Lb capacity
  • Delrin bearing
  • 5/16-18 x 1 in threaded stem
  • Hassle free guarantee