Learn More
MSC E.R. Wagner Stem Mount Swivel Caster, 2 in, Soft Rubber, 75 Lb Capacity
Catalog No. 5000533519
Encompass
This stem mount swivel caster features a 2 in diameter soft rubber wheel with a 75 lb capacity. It is designed with a threaded stem and a Delrin bearing, making it suitable for various furniture applications.
  • Threaded stem type
  • 3/8-16 stem diameter
  • Swivel functionality
  • 2 in wheel diameter
  • Soft rubber wheel material
  • Delrin bearing type
  • Hardened-heat treated construction
  • Soft wheel hardness
  • Chrome finish
  • Ideal for furniture casters
Catalog No. 50-005-33519 Supplier MSC Supplier No. 01504901
This stem mount swivel caster features a 2 in diameter soft rubber wheel with a 75 lb capacity. It is designed with a threaded stem and a Delrin bearing, making it suitable for various furniture applications.
  • Threaded stem type
  • 3/8-16 stem diameter
  • Swivel functionality
  • 2 in wheel diameter
  • Soft rubber wheel material
  • Delrin bearing type
  • Hardened-heat treated construction
  • Soft wheel hardness
  • Chrome finish
  • Ideal for furniture casters