Learn More
MSC E.R. Wagner Stem Mount Swivel Caster, 5 in, 6-1/8 in OAH, Soft Rubber, Friction Stem
Catalog No. 5000513219
Encompass
This stem mount swivel caster is designed for a range of applications, featuring a soft rubber wheel and a 200 Lb capacity. Its durable, hardened-heat treated construction and zinc finish make it suitable for use on hard surfaces, while also meeting NSF standards.
  • 5 inch diameter wheel
  • 1-1/4 inch wide wheel
  • 6-1/8 inch overall height
  • 200 Lb capacity
  • Delrin bearing for smooth operation
  • 7/16 inch friction stem
  • Zinc finish for corrosion resistance
  • Hardened-heat treated construction for durability
  • Suitable for hard surfaces
  • NSF standards included
  • Soft rubber wheel material
  • Swivel capability for maneuverability
Catalog No. 50-005-13219 Supplier MSC Supplier No. 01506310
This stem mount swivel caster is designed for a range of applications, featuring a soft rubber wheel and a 200 Lb capacity. Its durable, hardened-heat treated construction and zinc finish make it suitable for use on hard surfaces, while also meeting NSF standards.
  • 5 inch diameter wheel
  • 1-1/4 inch wide wheel
  • 6-1/8 inch overall height
  • 200 Lb capacity
  • Delrin bearing for smooth operation
  • 7/16 inch friction stem
  • Zinc finish for corrosion resistance
  • Hardened-heat treated construction for durability
  • Suitable for hard surfaces
  • NSF standards included
  • Soft rubber wheel material
  • Swivel capability for maneuverability