Learn More
MSC E.R. Wagner Stem Mount Swivel Caster, 5 inch, Polyolefin, Delrin Bearing, NSF
Catalog No. 5000513516
Encompass
This E.R. Wagner stem mount swivel caster is designed for various applications, featuring a polyolefin wheel, Delrin bearing, and NSF certification for reliable performance and durability.
  • 5 inch wheel diameter and 1-3/8 inch wheel width provide stable movement.
  • Polyolefin wheel material ensures durability and smooth operation.
  • Delrin bearing offers reduced friction and extended life.
  • Swivel design allows for easy maneuverability.
  • Zinc finish provides enhanced corrosion resistance.
  • Hardened-heat treated construction ensures robust performance.
  • Suitable for carpeted surfaces.
  • NSF certified for use in hygiene-sensitive environments.
  • 300 lb capacity for heavy-duty applications.
  • Features a 7/16 x 1-29/32 inch friction stem for secure mounting.
Catalog No. 50-005-13516 Supplier MSC Supplier No. 01506674
This E.R. Wagner stem mount swivel caster is designed for various applications, featuring a polyolefin wheel, Delrin bearing, and NSF certification for reliable performance and durability.
  • 5 inch wheel diameter and 1-3/8 inch wheel width provide stable movement.
  • Polyolefin wheel material ensures durability and smooth operation.
  • Delrin bearing offers reduced friction and extended life.
  • Swivel design allows for easy maneuverability.
  • Zinc finish provides enhanced corrosion resistance.
  • Hardened-heat treated construction ensures robust performance.
  • Suitable for carpeted surfaces.
  • NSF certified for use in hygiene-sensitive environments.
  • 300 lb capacity for heavy-duty applications.
  • Features a 7/16 x 1-29/32 inch friction stem for secure mounting.