Learn More
MSC E.R. Wagner Stem Mount Swivel Caster, Hard Rubber, 2-1/2 in, 115 lb, Delrin Bearing
Catalog No. 5000529834
Encompass
This stem mount swivel caster features a durable hard rubber wheel and is designed for applications requiring mobility and load support. It has a 2-1/2 in diameter, 115 lb capacity, and is equipped with a Delrin bearing. The caster utilizes a 3/8-16 x 1-1/2 in threaded stem for secure mounting.
  • Swivel functionality for enhanced maneuverability.
  • Durable hard rubber wheel material ensures longevity.
  • Equipped with a Delrin bearing for smooth operation.
  • Features a threaded stem for secure and easy mounting.
  • Hardened and heat-treated construction.
  • Chrome finish for corrosion resistance.
Catalog No. 50-005-29834 Supplier MSC Supplier No. 01505064
This stem mount swivel caster features a durable hard rubber wheel and is designed for applications requiring mobility and load support. It has a 2-1/2 in diameter, 115 lb capacity, and is equipped with a Delrin bearing. The caster utilizes a 3/8-16 x 1-1/2 in threaded stem for secure mounting.
  • Swivel functionality for enhanced maneuverability.
  • Durable hard rubber wheel material ensures longevity.
  • Equipped with a Delrin bearing for smooth operation.
  • Features a threaded stem for secure and easy mounting.
  • Hardened and heat-treated construction.
  • Chrome finish for corrosion resistance.