Learn More
MSC E.R. Wagner Swivel Caster, 3 in, Polyolefin, Threaded Stem
Catalog No. 5000529597
Encompass
This swivel caster is designed for industrial and commercial applications, featuring a stem mount for secure attachment. It includes a polyolefin wheel with a 3 inch diameter and 1-1/4 inch width, offering a 210 lb capacity with a plain bearing. The caster is equipped with a 7/16-14 x 1-1/2 inch threaded stem for easy installation.
  • Threaded stem mount for secure installation.
  • Wheel made from durable polyolefin material.
  • Provides a 3 inch wheel diameter.
  • Features a swivel design for maneuverability.
  • Equipped with a plain bearing.
  • Constructed with hardened-heat treated components.
  • Finished with zinc chromate for corrosion resistance.
  • Suitable for industrial and commercial applications.
  • Manufactured in the United States.
Catalog No. 50-005-29597 Supplier MSC Supplier No. 01506146
This swivel caster is designed for industrial and commercial applications, featuring a stem mount for secure attachment. It includes a polyolefin wheel with a 3 inch diameter and 1-1/4 inch width, offering a 210 lb capacity with a plain bearing. The caster is equipped with a 7/16-14 x 1-1/2 inch threaded stem for easy installation.
  • Threaded stem mount for secure installation.
  • Wheel made from durable polyolefin material.
  • Provides a 3 inch wheel diameter.
  • Features a swivel design for maneuverability.
  • Equipped with a plain bearing.
  • Constructed with hardened-heat treated components.
  • Finished with zinc chromate for corrosion resistance.
  • Suitable for industrial and commercial applications.
  • Manufactured in the United States.