Learn More
MSC E.R. Wagner Swivel Top Plate Caster, 3 in Phenolic, 250 lb Capacity
Catalog No. 5000529671
Encompass
This swivel top plate caster is equipped with a 3 in phenolic wheel and is designed for a variety of applications, including food service. It offers a load capacity of 250 lb and features a durable zinc finish.
  • Plate mount type
  • 3 in wheel diameter
  • Phenolic wheel material
  • 1-1/4 in wheel width
  • Black wheel color
  • Plain bearing type
  • Hardened-heat treated construction
  • Plate size of 3-1/8 x 4-1/8 in
  • 3-3/4 in overall height
  • Zinc finish/coating
  • 250 lb load capacity
  • Suitable for food service applications
Catalog No. 50-005-29671 Supplier MSC Supplier No. 01507896
This swivel top plate caster is equipped with a 3 in phenolic wheel and is designed for a variety of applications, including food service. It offers a load capacity of 250 lb and features a durable zinc finish.
  • Plate mount type
  • 3 in wheel diameter
  • Phenolic wheel material
  • 1-1/4 in wheel width
  • Black wheel color
  • Plain bearing type
  • Hardened-heat treated construction
  • Plate size of 3-1/8 x 4-1/8 in
  • 3-3/4 in overall height
  • Zinc finish/coating
  • 250 lb load capacity
  • Suitable for food service applications