Learn More
MSC E.R. Wagner Swivel Top Plate Caster, Phenolic, 5 in Wheel Diameter, 2 in Wheel Width, 650 lb Capacity
Catalog No. 5000535264
Encompass
This swivel top plate caster features a phenolic wheel with a 5 inch diameter and 2 inch width, providing a 650 lb load capacity. With an overall height of 6-1/2 inches, it includes a total-lock brake with a thread guard for enhanced safety and durability. Constructed from stamp-formed chrome zinc, this caster is designed for industrial and institutional applications.
  • Phenolic wheel material
  • 5 inch wheel diameter and 2 inch wheel width
  • 650 lb load capacity
  • 6-1/2 inch overall height
  • Total-lock brake with thread guard
  • Roller bearing type
  • Stamp formed construction with chrome zinc finish
  • 4 x 4-1/2 inch plate size
  • Top plate mount and swivel style
  • Suitable for industrial and institutional applications
Catalog No. 50-005-35264 Supplier MSC Supplier No. 57078685
This swivel top plate caster features a phenolic wheel with a 5 inch diameter and 2 inch width, providing a 650 lb load capacity. With an overall height of 6-1/2 inches, it includes a total-lock brake with a thread guard for enhanced safety and durability. Constructed from stamp-formed chrome zinc, this caster is designed for industrial and institutional applications.
  • Phenolic wheel material
  • 5 inch wheel diameter and 2 inch wheel width
  • 650 lb load capacity
  • 6-1/2 inch overall height
  • Total-lock brake with thread guard
  • Roller bearing type
  • Stamp formed construction with chrome zinc finish
  • 4 x 4-1/2 inch plate size
  • Top plate mount and swivel style
  • Suitable for industrial and institutional applications