Learn More
MSC E.R. Wagner Swivel Top Plate Caster, Phenolic, 8 inch Wheel, 1200 lb Capacity, Zinc Finish
Catalog No. 5000516986
Encompass
This swivel top plate caster features a phenolic wheel with an 8-inch diameter and a 2-inch width. It offers a 1200 lb load capacity and is designed with a corrosion-resistant zinc finish, making it suitable for institutional and industrial applications.
  • Double row swivel construction
  • Corrosion-resistant
Catalog No. 50-005-16986 Supplier MSC Supplier No. 01508555
This swivel top plate caster features a phenolic wheel with an 8-inch diameter and a 2-inch width. It offers a 1200 lb load capacity and is designed with a corrosion-resistant zinc finish, making it suitable for institutional and industrial applications.
  • Double row swivel construction
  • Corrosion-resistant