Learn More
MSC E.R. Wagner Swivel Top Plate Caster, Polyolefin, 4 inch Wheel Dia, 255 lb Capacity, Side Brake
Catalog No. 5000513140
Encompass
The E.R. Wagner Swivel Top Plate Caster is a versatile and durable caster designed for various applications. It features a polyolefin wheel, a 4-inch diameter, and a 255 lb load capacity. Equipped with a side brake, it offers reliable stopping power. The caster has a zinc finish and is suitable for use on carpeted surfaces.
  • Polyolefin wheel material
  • 4 inch wheel diameter
  • 1.25 inch wheel width
  • 255 lb load capacity
  • Side brake for secure stopping
  • Zinc finish
  • Designed for carpeted surfaces
  • Swivel top plate mount type
  • Overall height of 4.6875 inches
Catalog No. 50-005-13140 Supplier MSC Supplier No. 01507276
The E.R. Wagner Swivel Top Plate Caster is a versatile and durable caster designed for various applications. It features a polyolefin wheel, a 4-inch diameter, and a 255 lb load capacity. Equipped with a side brake, it offers reliable stopping power. The caster has a zinc finish and is suitable for use on carpeted surfaces.
  • Polyolefin wheel material
  • 4 inch wheel diameter
  • 1.25 inch wheel width
  • 255 lb load capacity
  • Side brake for secure stopping
  • Zinc finish
  • Designed for carpeted surfaces
  • Swivel top plate mount type
  • Overall height of 4.6875 inches