Learn More
MSC E.R. Wagner Swivel Top Plate Caster, Polypropylene, 5 in, 450 lb
Catalog No. 5000514496
Encompass
This swivel top plate caster features a polypropylene wheel with a 5 inch diameter and a 450 lb load capacity. Designed for medium-duty applications, it offers durability and smooth maneuverability for various tasks.
  • Polypropylene wheel material
  • 5 in wheel diameter
  • 450 lb load capacity
  • Swivel top plate mount
  • Roller bearing type
  • Hardened-heat treated construction
  • Black wheel color
  • Zinc finish
  • Overall height of 6-3/8 in
  • Plate size of 3-3/4 in x 4-5/8 in
Catalog No. 50-005-14496 Supplier MSC Supplier No. 57078073
This swivel top plate caster features a polypropylene wheel with a 5 inch diameter and a 450 lb load capacity. Designed for medium-duty applications, it offers durability and smooth maneuverability for various tasks.
  • Polypropylene wheel material
  • 5 in wheel diameter
  • 450 lb load capacity
  • Swivel top plate mount
  • Roller bearing type
  • Hardened-heat treated construction
  • Black wheel color
  • Zinc finish
  • Overall height of 6-3/8 in
  • Plate size of 3-3/4 in x 4-5/8 in