Learn More
MSC E.R. Wagner Swivel Top Plate Caster, Polyurethane, 4 in Wheel, 275 lb Capacity, Side Brake
Catalog No. 5000513378
Encompass
This swivel top plate caster features a durable polyurethane wheel, designed for various applications. It offers a 4 in wheel diameter, 1-1/4 in wheel width, and a substantial 275 lb load capacity, ensuring reliable performance. Equipped with a side brake, it provides enhanced control, and its overall height is 4-11/16 in, with plate dimensions of 3.75 x 2.75 in for secure mounting.
  • Polyurethane wheel material for durability
  • 4 in wheel diameter
  • 1-1/4 in wheel width
  • 275 lb load capacity
  • Side brake for enhanced control
  • 4-11/16 in overall height
  • 3.75 in plate length
  • 2.75 in plate width
  • Delrin bearing type
  • Hardened-heat treated construction
  • Zinc finish for corrosion resistance
  • Ideal for carpeted surfaces
Catalog No. 50-005-13378 Supplier MSC Supplier No. 01507243
This swivel top plate caster features a durable polyurethane wheel, designed for various applications. It offers a 4 in wheel diameter, 1-1/4 in wheel width, and a substantial 275 lb load capacity, ensuring reliable performance. Equipped with a side brake, it provides enhanced control, and its overall height is 4-11/16 in, with plate dimensions of 3.75 x 2.75 in for secure mounting.
  • Polyurethane wheel material for durability
  • 4 in wheel diameter
  • 1-1/4 in wheel width
  • 275 lb load capacity
  • Side brake for enhanced control
  • 4-11/16 in overall height
  • 3.75 in plate length
  • 2.75 in plate width
  • Delrin bearing type
  • Hardened-heat treated construction
  • Zinc finish for corrosion resistance
  • Ideal for carpeted surfaces