Learn More
MSC E.R. Wagner Swivel Top Plate Caster, Soft Rubber, 4 in
Catalog No. 5000514748
Encompass
This swivel top plate caster features a soft rubber wheel, designed for medium-duty applications. It offers a 4-inch wheel diameter, 1-1/4-inch wheel width, and a load capacity of 165 lbs. Equipped with a strap brake and a Delrin bearing, this caster ensures smooth movement and secure positioning. Its nickel finish provides durability, and it is NSF certified for various environments.
  • Soft rubber wheel for smooth operation.
  • 4-inch wheel diameter and 1-1/4-inch wheel width.
  • 165 lb load capacity for medium-duty applications.
  • Swivel top plate mount for maneuverability.
  • Strap brake for secure locking.
  • Delrin bearing for enhanced performance.
  • NSF certified, suitable for diverse uses.
  • Durable nickel finish.
Catalog No. 50-005-14748 Supplier MSC Supplier No. 58337718
This swivel top plate caster features a soft rubber wheel, designed for medium-duty applications. It offers a 4-inch wheel diameter, 1-1/4-inch wheel width, and a load capacity of 165 lbs. Equipped with a strap brake and a Delrin bearing, this caster ensures smooth movement and secure positioning. Its nickel finish provides durability, and it is NSF certified for various environments.
  • Soft rubber wheel for smooth operation.
  • 4-inch wheel diameter and 1-1/4-inch wheel width.
  • 165 lb load capacity for medium-duty applications.
  • Swivel top plate mount for maneuverability.
  • Strap brake for secure locking.
  • Delrin bearing for enhanced performance.
  • NSF certified, suitable for diverse uses.
  • Durable nickel finish.